A2627
Adenosine 5′-monophosphoramidate sodium salt
~95%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H14N6NaO6P
CAS Number:
Molecular Weight:
368.22
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Assay
~95%
form
solid
storage temp.
−20°C
SMILES string
[Na+].Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(N)([O-])=O)[C@@H](O)[C@H]3O
InChI
1S/C10H15N6O6P.Na/c11-8-5-9(14-2-13-8)16(3-15-5)10-7(18)6(17)4(22-10)1-21-23(12,19)20;/h2-4,6-7,10,17-18H,1H2,(H2,11,13,14)(H3,12,19,20);/q;+1/p-1/t4-,6-,7-,10-;/m1./s1
InChI key
ALRHXZAMLVPXPL-MCDZGGTQSA-M
Biochem/physiol Actions
Intermediate for nucleotide synthesis
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service