Skip to Content
Merck
All Photos(1)

Key Documents

A1546

Supelco

N-(4-Aminobutyl)-5-chloro-1-naphthalenesulfonamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17ClN2O2S · HCl
CAS Number:
Molecular Weight:
349.28
MDL number:
UNSPSC Code:
41116107

storage temp.

2-8°C

SMILES string

Cl.NCCCCNS(=O)(=O)c1cccc2c(Cl)cccc12

Biochem/physiol Actions

Antitumor agent which inhibits blood platelet aggregation; a calmodulin antagonist

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Hidaka et al.
Proceedings of the National Academy of Sciences of the United States of America, 78(7), 4354-4357 (1981-07-01)
N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide (W-7) and its derivatives are putative calmodulin antagonists that bind to calmodulin and inhibit Ca2+/calmodulin-regulated enzyme activities. Autoradiographic studies using tritiated W-7 showed that this compound penetrates the cell membrane, is distributed mainly in the cytoplasm, and inhibits proliferation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service