All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H5N5
CAS Number:
Molecular Weight:
135.13
MDL number:
UNSPSC Code:
12352200
form
aqueous ethanol solution
extent of labeling
5-15 Ci/mmol
mass shift
M+4
storage temp.
2-8°C
SMILES string
[3H]c1nc(N)c2nc([3H])[nH]c2n1
Packaging
serum bottle
Physical form
Aqueous solution containing 2% ethanol
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service