Skip to Content
Merck
All Photos(1)

Documents

42232

Sigma-Aldrich

Nonyl-β-D-thioglucoside

≥99% (TLC)

Synonym(s):

n-Nonyl β-D-thioglucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H30O5S
CAS Number:
Molecular Weight:
322.46
Beilstein:
6058993
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (TLC)

form

solid

pH

5-8

conductivity

≤40 μS/cm (0.05% in methanol:water (1:1))

solubility

methanol: water (1:1): 0.05%, clear, colorless

absorption

≤0.1 at 340 nm
≤0.2 at 280 nm
≤0.25 at 260 nm

SMILES string

CCCCCCCCCS[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C15H30O5S/c1-2-3-4-5-6-7-8-9-21-15-14(19)13(18)12(17)11(10-16)20-15/h11-19H,2-10H2,1H3/t11-,12-,13+,14-,15+/m1/s1

InChI key

HICSQFRUQXWIGI-QMIVOQANSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryan W Berger et al.
Biophysical journal, 89(1), 452-464 (2005-04-26)
The effects of various surfactants on the activity and stability of the human adenosine A3 receptor (A3) were investigated. The receptor was expressed using stably transfected HEK293 cells at a concentration of 44 pmol functional receptor per milligram membrane protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service