Skip to Content
Merck
All Photos(1)

Documents

Y0000455

Propafenone hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-[2-(2-Hydroxy-3-(propylamino)propoxy)phenyl]-3-phenyl-1-propanone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27NO3 · HCl
CAS Number:
Molecular Weight:
377.90
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

propafenone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CCCNCC(O)COc1ccccc1C(=O)CCc2ccccc2

InChI

1S/C21H27NO3.ClH/c1-2-14-22-15-18(23)16-25-21-11-7-6-10-19(21)20(24)13-12-17-8-4-3-5-9-17;/h3-11,18,22-23H,2,12-16H2,1H3;1H

InChI key

XWIHRGFIPXWGEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Propafenone hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michela Faggioni et al.
Circulation. Arrhythmia and electrophysiology, 7(2), 313-320 (2014-02-05)
Atrial fibrillation (AF) risk has been associated with leaky ryanodine receptor 2 (RyR2) Ca release channels. Patients with mutations in RyR2 or in the sarcoplasmic reticulum Ca-binding protein calsequestrin 2 (Casq2) display an increased risk for AF. Here, we examine
Naveen Rajpurohit et al.
South Dakota medicine : the journal of the South Dakota State Medical Association, 67(4), 137-139 (2014-05-06)
We describe a rare case of severe drug-drug interaction between propafenone and mirtazapine leading to propafenone toxicity. A 69-year-old Caucasian male taking propafenone for atrial fibrillation was prescribed mirtazapine for insomnia. Subsequent to the first dose of mirtazapine the patient
Nancy M Allen LaPointe et al.
The American journal of cardiology, 115(3), 316-322 (2014-12-11)
Little is known in clinical practice about antiarrhythmic drug (AAD) use in patients with atrial fibrillation (AF) (particularly younger ones) who do not have structural heart disease. Using the MarketScan database, we identified patients <65 years without known coronary artery disease
Jean-François Desaphy et al.
Experimental neurology, 255, 96-102 (2014-03-13)
Although the sodium channel blocker mexiletine is considered the first-line drug in myotonia, some patients experiment adverse effects, while others do not gain any benefit. Other antimyotonic drugs are thus needed to offer mexiletine alternatives. In the present study, we
Hwa Yeon Yi et al.
The American journal of emergency medicine, 32(10), 1200-1207 (2014-08-26)
Unlike other sodium-channel-blocking antiarrhythmic agents, propafenone has β-blocking effects and calcium-channel-blocking effects. Yi et al recently studied insulin's treatment effect on acute propafenone toxicity in rats. However, because the degree of effectiveness of insulin compared to the previously known antidote

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service