Skip to Content
Merck
All Photos(1)

Documents

F1756

Sigma-Aldrich

6-[Fluorescein-5(6)-carboxamido]hexanoic acid N-hydroxysuccinimide ester

Technical grade

Synonym(s):

Fluorescein-5(6)-carboxamidocaproic acid N-succinimidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H26N2O10
CAS Number:
Molecular Weight:
586.55
MDL number:
UNSPSC Code:
12352200

storage temp.

−20°C

SMILES string

Oc1ccc2c(Oc3cc(O)ccc3C24OC(=O)c5cc(ccc45)C(=O)NCCCCCC(=O)ON6C(=O)CCC6=O)c1.Oc7ccc8c(Oc9cc(O)ccc9C8%10OC(=O)c%11ccc(cc%10%11)C(=O)NCCCCCC(=O)ON%12C(=O)CCC%12=O)c7

InChI

1S/2C31H26N2O10/c34-18-6-9-22-24(15-18)41-25-16-19(35)7-10-23(25)31(22)21-8-5-17(14-20(21)30(40)42-31)29(39)32-13-3-1-2-4-28(38)43-33-26(36)11-12-27(33)37;34-18-6-9-21-24(15-18)41-25-16-19(35)7-10-22(25)31(21)23-14-17(5-8-20(23)30(40)42-31)29(39)32-13-3-1-2-4-28(38)43-33-26(36)11-12-27(33)37/h2*5-10,14-16,34-35H,1-4,11-13H2,(H,32,39)

InChI key

WAGYELYAQOYZFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for introducing the fluorescein label; the spacer renders the reactive group more accessible to nucleophiles on biopolymers

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service