Skip to Content
Merck
All Photos(1)

Key Documents

93191

Sigma-Aldrich

Triphenyltin chloride

purum, ≥97.0% (AT)

Synonym(s):

Chlorotriphenylstannane, Fentin chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3SnCl
CAS Number:
Molecular Weight:
385.47
Beilstein:
524762
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

purum

Assay

≥97.0% (AT)

bp

240 °C/13.5 mmHg (lit.)

mp

103-106 °C
108 °C (dec.) (lit.)

SMILES string

Cl[Sn](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/3C6H5.ClH.Sn/c3*1-2-4-6-5-3-1;;/h3*1-5H;1H;/q;;;;+1/p-1

InChI key

NJVOZLGKTAPUTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>302.0 °F

Flash Point(C)

> 150 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guanyong Su et al.
Environmental science and pollution research international, 20(2), 803-811 (2012-11-07)
Triphenyltin chloride (TPTC), which has been extensively used in industry and agriculture, can occur at concentrations in the environment sufficient to be toxic. Here, potency of TPTC to modulate genes in a library containing 1,820 modified green fluorescent protein (GFP)-expressing
Bao-an Wang et al.
Zhonghua nan ke xue = National journal of andrology, 12(6), 516-519 (2006-07-13)
To investigate the effects of tributyltin chloride (TBT) and triphenyltin chloride (TPT) on rat testicular Leydig cells. The rat Leydig cells (LC-540) were incubated with 0 to 80 nmol/L TBT and TPT for 24 to approximately 96 h, and then
Angeliki Lyssimachou et al.
Aquatic toxicology (Amsterdam, Netherlands), 89(2), 129-135 (2008-07-25)
Long-term exposures to organotin compounds have shown alterations on endogenous steroid levels in gastropods together with the development of imposex. However, information regarding short-term effects of these compounds on the endocrine system of gastropods is lacking. This work aimed at
Konstanze Grote et al.
Toxicology, 260(1-3), 53-59 (2009-05-26)
Consumers are exposed to organotin compounds (OTCs) via contaminated fish and seafood due to the accumulation of these compounds in marine organisms. Certain OTCs are immunotoxic and may also have endocrine disrupting properties resulting in adverse effects on the reproductive
Eun-Min Cho et al.
Journal of microbiology and biotechnology, 22(3), 378-384 (2012-03-28)
In aquatic invertebrates, particularly marine gastropods, organotin compounds induce irreversible sexual abnormality in females, which is termed imposex, at very low concentrations. Organotin compounds are agonists for nuclear receptors such as RXRs and PPARgamma. However, the imposex phenomenon has not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service