Skip to Content
Merck
All Photos(1)

Key Documents

92890

Sigma-Aldrich

1,3,5-Trioxane

purum, ≥99.0% (GC)

Synonym(s):

Metaformaldehyde, Trioxymethylene, sym.-Trioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
Beilstein:
102769
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

purum

Assay

≥99.0% (GC)

autoignition temp.

777 °F

expl. lim.

29 %

bp

112-115 °C

mp

59-62 °C (lit.)
59-62 °C

SMILES string

C1OCOCO1

InChI

1S/C3H6O3/c1-4-2-6-3-5-1/h1-3H2

InChI key

BGJSXRVXTHVRSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Barański et al.
Medycyna pracy, 35(4), 245-255 (1984-01-01)
The study was aimed at evaluation of the effects of trioxane and dioxolane on the fertility and frequency of dominant lethal mutations in germ cells of male rats. In the first experiment each test compound was administered per os at
Paweł Kaszycki et al.
Biodegradation, 13(2), 91-99 (2002-11-27)
Methylotrophic yeast Hansenula polymorpha were shown to cooperate with activated sludge from biological wastewater treatment stations, enhancing substantially its potential to biodegrade formaldehyde in industrial wastewater. After integration with yeast cells the modified sludge retained its original structure and activity
T Czajkowska et al.
Medycyna pracy, 38(4), 244-249 (1987-01-01)
The studies on the accumulation of toxic effects of TOX and DOX have been carried out with the Kagan and Stańkiewicz method. For 7 months the rats were intragastrically administered, with a tube, 20% water solutions of TOX and DOX
K Sitarek et al.
Polish journal of occupational medicine, 3(2), 209-213 (1990-01-01)
An aqueous solution of trioxane was administered by gavage to female rats, 5 days per week for 7 weeks, at doses of 0.19, 0.58 and 1.16 g/kg/day. A significant increase in the mean duration of the oestrous cycle, mainly due
Z Kowalski et al.
Mutation research, 136(3), 169-171 (1984-06-01)
The mutagenic activity of trioxane and dioxolane was investigated in 5 histidine-requiring strains of S. typhimurium: TA1535, TA1537, TA1538, TA98 and TA100 with and without activation by liver microsomes. They were not mutagenic under any of these conditions with any

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service