Skip to Content
Merck
All Photos(1)

Key Documents

92661

Supelco

Sebacoyl-L-carnitine-(N-methyl-d3) lithium salt

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17D3H28NO6 · xLi+
Molecular Weight:
348.45 (free acid basis)
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

form

solid

optical activity

[α]/D -15.0±2.0°, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

extent of labeling

≥99.0%

impurities

1.5-3.0% lithium

application(s)

clinical testing

storage temp.

−20°C

SMILES string

C[N+](C[C@H](OC(CCCCCCCCC(O)=O)=O)CC([O-])=O)(C)C([2H])([2H])[2H]

InChI key

GBFPILOKXGQZKW-XEVIKCSCSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service