Skip to Content
Merck
All Photos(1)

Documents

91277

Supelco

1,3-Bis(2-methoxyphenoxy)-2-propanol

pharmaceutical impurity standard

Synonym(s):

Guaifenesin impurity D (PhEur), 1,3-Bis(2-methoxyphenoxy)propan-2-ol, NSC 142122

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20O5
CAS Number:
Molecular Weight:
304.34
Beilstein:
2003232
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(COC1=C(OC)C=CC=C1)COC2=CC=CC=C2OC

InChI

1S/C17H20O5/c1-19-14-7-3-5-9-16(14)21-11-13(18)12-22-17-10-6-4-8-15(17)20-2/h3-10,13,18H,11-12H2,1-2H3

InChI key

SSNCGCIXFDVLFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service