Skip to Content
Merck
All Photos(2)

Documents

907456

Sigma-Aldrich

(2R,4aR,6R,7S,8R,8aS)-2-Phenyl-6-(phenylthio)hexahydropyrano[3,2-d][1,3]dioxine-7,8-diol

≥95%

Synonym(s):

Phenyl 4,6-O-benzylidine-1-thio-α-D-mannoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20O5S
CAS Number:
Molecular Weight:
360.42
MDL number:
UNSPSC Code:
12352005

Assay

≥95%

form

powder

mp

210-212 °C

storage temp.

2-8°C

InChI

1S/C19H20O5S/c20-15-16(21)19(25-13-9-5-2-6-10-13)23-14-11-22-18(24-17(14)15)12-7-3-1-4-8-12/h1-10,14-21H,11H2/t14?,15?,16-,17-,18?,19-/m1/s1

InChI key

BDNIQCYVYFGHSI-FCUBMCITSA-N

Application

This is a protected mannopyranoside useful as a building block for synthesis of complex carbohydrates. The compound has α-phenylthio and 4,6-benzylidene protecting groups

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Carbohydrates and their biologically active glycoconjugate analogs are emerging as an important class of biomolecules.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service