Skip to Content
Merck
All Photos(1)

Key Documents

78361

Sigma-Aldrich

Fluorescent red 630 reactive

for fluorescence, ≥80% (coupling to amines)

Synonym(s):

Reactive fluorescent red 630

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H49N3O8S
CAS Number:
Molecular Weight:
731.90
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

for fluorescence

Assay

≥80% (coupling to amines)

solubility

DMF: soluble

fluorescence

λex 620 nm; λem 650 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CCN(CC)c1ccc2c(\C=C\C=C3\N(CCCCCC(=O)ON4C(=O)CCC4=O)c5ccc(cc5C3(C)C)S([O-])(=O)=O)cc([o+]c2c1)C(C)(C)C

InChI

1S/C40H49N3O8S/c1-8-41(9-2)28-17-19-30-27(24-35(39(3,4)5)50-33(30)25-28)14-13-15-34-40(6,7)31-26-29(52(47,48)49)18-20-32(31)42(34)23-12-10-11-16-38(46)51-43-36(44)21-22-37(43)45/h13-15,17-20,24-26H,8-12,16,21-23H2,1-7H3/b14-13+,34-15+

InChI key

PHHUECZJTSYKRH-DBYPPFPASA-N

Application

Amino-reactive biolabel excited by red lasers (He/Ne; 633 nm) and diode lasers (635-650 nm). The chromophore is a bridged hemicyanine. Quantum yield is fairly low in aqueous media, but increases up to one order of magnitude if coupled to a target protein; Molar absorption 120,000 M-1 cm-1, λabs: (conjugate) 645 nm; λem: (conjugate) 660 nm

Features and Benefits

This novel hemocyanin dye is optimally excited by He-Ne laser with a wavelength of 633 nm, but may also be excited by argon or argon-krypton laser with a wavelength of 647 nm or diode lasers.
Additional advantages are:
High molar absorptions (120,000 M-1cm-1)
Low quantum yields in nonaqueous media, greatly increased by binding to target molecules.
Very strong fluorescence in solid state, which allows preparation of DNA for arrays on glass.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service