Skip to Content
Merck
All Photos(1)

Key Documents

69369

Supelco

3-Methylmalic acid

≥95.0% (HPLC)

Synonym(s):

β-Methylmalic acid, (±)-2-Hydroxy-3-methylbutandioic acid, 2-Hydroxy-3-methylsuccinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O5
CAS Number:
Molecular Weight:
148.11
Beilstein:
1773280
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

Assay

≥95.0% (HPLC)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC(C(C)C(O)C(O)=O)=O

InChI

1S/C5H8O5/c1-2(4(7)8)3(6)5(9)10/h2-3,6H,1H3,(H,7,8)(H,9,10)

InChI key

NPYQJIHHTGFBLN-UHFFFAOYSA-N

Biochem/physiol Actions

Metabolite of C5-branched dibasic acid metabolism, valine, leucine and isoleucine biosynthesis and 2-oxocarboxylic acid metabolism.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sylvia Herter et al.
The Journal of biological chemistry, 277(23), 20277-20283 (2002-04-04)
Phototrophic CO(2) assimilation by the primitive, green eubacterium Chloroflexus aurantiacus has been shown earlier to proceed in a cyclic mode via 3-hydroxypropionate, propionyl-CoA, succinyl-CoA, and malyl-CoA. The metabolic cycle could be closed by cleavage of malyl-CoA affording glyoxylate (the primary
Juliane Elisa Welke et al.
Journal of chromatography. A, 1226, 124-139 (2012-01-27)
Wine aroma is an important characteristic and may be related to certain specific parameters, such as raw material and production process. The complexity of Merlot wine aroma was considered suitable for comprehensive two-dimensional gas chromatography (GC×GC), as this technique offers
D T Loots
Journal of inherited metabolic disease, 32(3), 403-411 (2009-04-04)
Although a number of abnormal diagnostic metabolites have previously been described in the urine of patients with isovaleric acidaemia (IVA), they do not fully explain the clinical symptoms associated with this disease. On the basis of our current understanding of
I Roymoulik et al.
Biochemistry, 39(33), 10340-10346 (2000-08-24)
Adenosylcobalamin-dependent enzymes catalyze a variety of chemically difficult isomerizations in which a nonacidic hydrogen on one carbon is interchanged with an electron-withdrawing group on an adjacent carbon. We describe a new isomerization, that of L-2-hydroxyglutarate to L-threo-3-methylmalate, involving the migration
Laura Strittmatter et al.
Human molecular genetics, 23(9), 2313-2323 (2013-12-18)
CLYBL is a human mitochondrial enzyme of unknown function that is found in multiple eukaryotic taxa and conserved to bacteria. The protein is expressed in the mitochondria of all mammalian organs, with highest expression in brown fat and kidney. Approximately

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service