Skip to Content
Merck
All Photos(1)

Documents

65371

Supelco

(±)-α-Methoxy-α-trifluoromethylphenylacetic acid

for GC derivatization, LiChropur, ≥97.0% (T)

Synonym(s):

MTPA, Mosher′s acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9F3O3
CAS Number:
Molecular Weight:
234.17
Beilstein:
2215439
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥97.0% (T)

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

mp

40-45 °C

storage temp.

2-8°C

SMILES string

COC(C(O)=O)(c1ccccc1)C(F)(F)F

InChI

1S/C10H9F3O3/c1-16-9(8(14)15,10(11,12)13)7-5-3-2-4-6-7/h2-6H,1H3,(H,14,15)

InChI key

JJYKJUXBWFATTE-UHFFFAOYSA-N

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

a-Methoxy-a-trifluoromethylphenylacetic acid, a versatile reagent for the determination of enantiomeric composition of alcohols and amines.
J. A. Dale, D. L. Dull, H. S. Mosher
The Journal of Organic Chemistry, 34(9), 2543-2549 (1969)
Dongsheng Du et al.
Fitoterapia, 101, 73-79 (2015-01-07)
Two new germacrane sesquiterpenes, yedoensins A (1) and B (2), together with 8 known ones (3-10) were isolated from the herb of Viola yedoensis. The structures of the new compounds were established by extensive spectroscopic means including 1D ((1)H and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service