Skip to Content
Merck
All Photos(1)

Key Documents

53603

Supelco

N-Adipoylglycine

analytical standard

Synonym(s):

6-[(Carboxymethyl)amino]-6-oxohexanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13NO5
CAS Number:
Molecular Weight:
203.19
Beilstein:
2265543
MDL number:
UNSPSC Code:
12352200

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

OC(CNC(CCCCC(O)=O)=O)=O

InChI

1S/C8H13NO5/c10-6(9-5-8(13)14)3-1-2-4-7(11)12/h1-5H2,(H,9,10)(H,11,12)(H,13,14)

InChI key

UJHDYVYBNDNRRQ-UHFFFAOYSA-N

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service