Skip to Content
Merck
All Photos(1)

Documents

513849

Sigma-Aldrich

Thymolphthalein Complexone sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H44N2Na2O12
CAS Number:
Molecular Weight:
766.74
MDL number:
UNSPSC Code:
12171500

mp

209 °C (dec.) (lit.)

SMILES string

[Na].[Na].CC(C)c1cc(c(C)c(CN(CC(O)=O)CC(O)=O)c1O)C2(OC(=O)c3ccccc23)c4cc(C(C)C)c(O)c(CN(CC(O)=O)CC(O)=O)c4C

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service