Skip to Content
Merck
All Photos(1)

Key Documents

44184

Supelco

Pymetrozin

reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N5O
CAS Number:
Molecular Weight:
217.23
Beilstein:
7814151
MDL number:
UNSPSC Code:
41116107

grade

reference material

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

CC1=NNC(=O)N(C1)\N=C\c2cccnc2

InChI

1S/C10H11N5O/c1-8-7-15(10(16)14-13-8)12-6-9-3-2-4-11-5-9/h2-6H,7H2,1H3,(H,14,16)/b12-6+

InChI key

QHMTXANCGGJZRX-WUXMJOGZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Xiang Wang et al.
Pesticide biochemistry and physiology, 165, 104548-104548 (2020-05-04)
Pymetrozine is a promising chemical used to control brown planthopper, which developed resistance to imidacloprid and buprofezin in the past decade. Field efficacy indicates that pymetrozine can reduce the number of offsprings of brown planthopper, but the specific physiological mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service