Skip to Content
Merck
All Photos(1)

Key Documents

43226

Supelco

Fenchyl alcohol

analytical standard

Synonym(s):

Fenchol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

mp

39.0-45.0 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H][C@]12CC[C@](C)(C1)[C@@H](O)C2(C)C

InChI

1S/C10H18O/c1-9(2)7-4-5-10(3,6-7)8(9)11/h7-8,11H,4-6H2,1-3H3/t7-,8-,10+/m0/s1

InChI key

IAIHUHQCLTYTSF-OYNCUSHFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fenchyl alcohol is a fragrance ingredient used in certain cosmetic products and non-cosmetic products such as household cleaners and detergents.

Application

Fenchyl alcohol may be used as a model compound to determine the effective concentration of volatile flavor compounds in aqueous model systems by reversed-phase high-performance liquid chromatography method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fragrance material review on fenchyl alcohol.
Bhatia S P, et al.
Food And Chemical Toxicology, 46(11), S157-S159 (2008)
A simple method for concentrating volatile flavor compounds in aqueous systems using RP-HPLC.
Aishima T, et al.
Chromatographia, 28(7-8), 405-411 (1989)
S P Bhatia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 11, S157-S159 (2008-07-22)
A toxicologic and dermatologic review of fenchyl alcohol when used as a fragrance ingredient is presented.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service