Skip to Content
Merck
All Photos(1)

Documents

40538

Supelco

Pinnatoxin F solution

20 μg/g (in 0.1% acetic acid in 50% acetonitrile, nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

20 μg/g (in 0.1% acetic acid in 50% acetonitrile, nominal concentration)

format

single component solution

storage temp.

−20°C

SMILES string

C=C(C[C@H]1O[C@]2(O[C@@]3(O[C@@H]4[C@@H](O)[C@@H](C)C3)CC2)[C@](C)(O)CC1)CCCC5=NC[C@H](C)[C@@H](C)C[C@]65CCC(C7OC(CC7)=O)=C[C@]6([H])[C@@H]8O[C@]9(C4)C[C@@H](C)C[C@H]8O9

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI.
Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

For the analysis of marine toxins in seafood.

Check out our complete range of marine toxins reference materials

Analysis Note

20 μg/g in 0.1% acetic acid in 50% acetonitrile

Other Notes

Dihydro-5-[(2S,3R,11S,14R,15R,18S,20S,21S,22S,24S,26S,28R,29S,29aS,33aR)-2,3,4,6,7,8,9,10,11,12,13,14,16,17,20,21,25,26,27,28,29,29a,32,33-tetracosahydro-14,21-dihydroxy-2,3,14,20,26-pentamethyl-9-methylene-19H-11,15:15,18:18,22:24,28:24,29-pentaepoxy-1H-benzo[2,3]cycloheptacos[1,2-b]azepin-31-yl]-2(3H)-furanone

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service