Skip to Content
Merck
All Photos(1)

Key Documents

37404

Supelco

AQUANAL-plus iron (Fe) 0.2-15 mg/L

test set with color comparator and check solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41113319

usage

sufficient for 500 tests

measuring range

0.1-15 mg/L (photometric, Spectro 3)
0.2-15 mg/L (color chart)

application(s)

environmental
general analytical

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Method
Formation of a red color complex with 1,10-phenanthroline

Principle
DEV

Gradation (color chart)
0 - 0.2 - 0.5 - 1.0 - 2.0 - 3.0 - 5.0 - 7.5 - 10 - 15 mg/L (Fe2+/3+)

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service