Skip to Content
Merck
All Photos(1)

Key Documents

36500

Supelco

Dialifos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17ClNO4PS2
CAS Number:
Molecular Weight:
393.85
Beilstein:
1550800
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)SC(CCl)N1C(=O)c2ccccc2C1=O

InChI

1S/C14H17ClNO4PS2/c1-3-19-21(22,20-4-2)23-12(9-15)16-13(17)10-7-5-6-8-11(10)14(16)18/h5-8,12H,3-4,9H2,1-2H3

InChI key

MUMQYXACQUZOFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Environmental fate of dialifor and formation of its oxygen analogue following application on grape vines in the San Joaquin Valley, California.
W Winterlin et al.
Journal of agricultural and food chemistry, 28(6), 1078-1083 (1980-11-01)
M Kalafatić et al.
The International journal of developmental biology, 35(3), 335-340 (1991-09-01)
Investigations showed that the three insecticides used had the most damaging effect upon hydra immediately after treatment. The tentacles and the hypostome are the parts most often damaged. Inse the affected cells, lesions appear in the intracellular membranes, the nucleus
R Besser et al.
Neurology, 40(8), 1275-1277 (1990-08-01)
Two patients with acute severe organophosphate intoxication showed (1) single evoked compound muscle action potentials (CMAP) with repetitive discharges and (2) prominent decremental responses of CMAP with 20 and 50 Hz supramaximal nerve stimulation. Following the intravenous injection of single
Prashant Shankarrao Adole et al.
Analytical biochemistry, 621, 114158-114158 (2021-03-12)
Intentional or unintentional intake of anticholinesterase pesticides became common due to their extensive use in agricultural and domestic purposes, resulting in numerous poisoning cases. A simple, accurate, and sensitive gas chromatography-ion trap mass spectrometry-based method for the quantification of 12
H N Nigg et al.
Archives of environmental contamination and toxicology, 10(4), 497-504 (1981-07-01)
Surface residue disappearance rates of dioxathion, malathion, oxydemetonmethyl, and dialifor were the same for fruit and leaves, and they increase with temperature. Disappearance rates were in the order of malathion greater than or equal to oxydemetonmethyl greater than dialifor greater

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service