Skip to Content
Merck
All Photos(1)

Documents

32525

Supelco

Flumioxazin

PESTANAL®, analytical standard

Synonym(s):

2-[7-Fluoro-3,4-dihydro-3-oxo-4-(2-propynyl)-2H-1,4-benzoxazin-6-yl]-4,5,6,7-tetrahydro-1H-isoindole-1,3(2H)-dione, Flumioxazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15FN2O4
CAS Number:
Molecular Weight:
354.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Fc1cc2OCC(=O)N(CC#C)c2cc1N3C(=O)C4=C(CCCC4)C3=O

InChI

1S/C19H15FN2O4/c1-2-7-21-15-9-14(13(20)8-16(15)26-10-17(21)23)22-18(24)11-5-3-4-6-12(11)19(22)25/h1,8-9H,3-7,10H2

InChI key

FOUWCSDKDDHKQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flumioxazin is an herbicide, belonging to the family of Benzoxazinones, that are organic compounds containing benzene fused to an oxazine ring (a six-member aliphatic ring with four carbon atoms, one oxygen atom, and one nitrogen atom) bearing a ketone group. It is used on soybeans, peanuts, orchard fruits and other crops.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lia Marchi-Werle et al.
Journal of economic entomology, 110(5), 2100-2108 (2017-09-30)
The soybean aphid, Aphis glycines Matsumura (Hemiptera: Aphididae), is an invasive species from Asia that has been the major economic insect pest of soybeans, Glycine max (L.) Merrill, since 2000. While use of soybeans expressing antibiosis and antixenosis is a
James T Brosnan et al.
Pest management science, 76(6), 2049-2057 (2020-01-17)
Indaziflam is an alkylazine herbicide used to control annual bluegrass (Poa annua L.). Several locations in the southeastern USA reported poor annual bluegrass control following treatment with indaziflam during autumn 2015. A series of controlled environment experiments were conducted to
Ethann R Barnes et al.
Frontiers in plant science, 8, 1455-1455 (2017-09-05)
Common ragweed emerges early in the season in Nebraska, USA and is competitive with soybean; therefore, preplant herbicides are important for effective control. Glyphosate has been used as a preplant control option; however, confirmation of glyphosate-resistant (GR) common ragweed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service