Skip to Content
Merck
All Photos(1)

Key Documents

31013

Sigma-Aldrich

Mercury(II) sulfate

puriss. p.a., ACS reagent, ≥99%

Synonym(s):

Mercuric sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgSO4
CAS Number:
Molecular Weight:
296.65
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

Assay

≥99%

ign. residue (after red. with HCOOH)

≤0.02% (as SO4)

anion traces

chloride (Cl-): ≤30 mg/kg
nitrate (NO3-): ≤30 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤1 mg/kg
Cu: ≤5 mg/kg
Fe: ≤30 mg/kg
Hg+: ≤500 mg/kg
K: ≤20 mg/kg
Mg: ≤30 mg/kg
Na: ≤50 mg/kg
Pb: ≤5 mg/kg

SMILES string

[Hg++].[O-]S([O-])(=O)=O

InChI

1S/Hg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

DOBUSJIVSSJEDA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L A Ivanova et al.
Gigiena i sanitariia, (11)(11), 66-67 (1991-11-01)
In experiments with two tissue culture lines it was shown, that cytotoxicity of mercury (1) derivatives (measured by CL50) is lesser two times, than it was for mercury (2) derivates. No significant differences in cytotoxicity were noted by the cell
A P de Souza et al.
Dental materials : official publication of the Academy of Dental Materials, 16(2), 103-108 (2001-02-24)
The interaction between metal ions and the oral environment is a major subject matter in dental research. Matrix metalloproteinases (MMPs) have been implicated in several pathologic oral processes such as periodontal tissue destruction, root caries, tumour invasion and temporomandibular joint
Paul I Dargan et al.
Critical care (London, England), 7(3), R1-R6 (2003-06-10)
Inorganic mercury poisoning is uncommon, but when it occurs it can result in severe, life-threatening features and acute renal failure. Previous reports on the use of extracorporeal procedures such as haemodialysis and haemoperfusion have shown no significant removal of mercury.
Necati Kayaalp et al.
Environmental science and pollution research international, 17(9), 1547-1552 (2010-05-25)
Chemical oxygen demand (COD) is used as a discharge standard parameter in wastewater treatment plant design, environmental modelling and many other applications. Chloride interference is an important problem of COD measurement for wastewaters containing low organic matter and high chloride
[Poisoning in treating scabies with mercuric sulfate ointment].
B G Belkin
Vestnik dermatologii i venerologii, (12)(12), 44-46 (1986-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service