26710-U
Hamilton® needles for RN syringes, blunt tip
volume 250-10000 μL, needle size 16 ga (blunt tip), blunt tip (PEEK), needle L 51 mm (2 in.)
About This Item
Recommended Products
material
blunt tip PEEK needle (point style 3)
needle size
16 ga (blunt tip)
description
replacement needle
packaging
pkg of 6 ea
manufacturer/tradename
Hamilton 8650-01
needle L
51 mm (2 in.)
volume
250-10000 μL
compatibility
for use with Hamilton 250μL-10mL syringe
for use with Rheodyne Injection Valves
Looking for similar products? Visit Product Comparison Guide
Footnote
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service