Skip to Content
Merck
All Photos(1)

Key Documents

13651

Sigma-Aldrich

Nickel(II) sulfate hexahydrate

puriss., ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiSO4 · 6H2O
CAS Number:
Molecular Weight:
262.85
EC Number:
MDL number:
UNSPSC Code:
12352300

vapor density

2.07 (vs air)

grade

puriss.

Assay

≥99%

pH

4-6 (20 °C, 5%)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

As: ≤10 mg/kg
Cd: ≤50 mg/kg
Co: ≤100 mg/kg
Cu: ≤20 mg/kg
Fe: ≤50 mg/kg
Pb: ≤20 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

InChI

1S/Ni.H2O4S.6H2O/c;1-5(2,3)4;;;;;;/h;(H2,1,2,3,4);6*1H2/q+2;;;;;;;/p-2

InChI key

RRIWRJBSCGCBID-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonia A Nemec et al.
Toxicological sciences : an official journal of the Society of Toxicology, 110(1), 212-223 (2009-05-01)
Hexavalent chromium (Cr(VI)) promotes lung injury and pulmonary diseases through poorly defined mechanisms that may involve the silencing of inducible protective genes. The current study investigated the hypothesis that Cr(VI) actively signals through a signal transducer and activator of transcription

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service