Skip to Content
Merck
All Photos(1)

Key Documents

11353

Supelco

1,8-Anthracenedimethanamine

≥98.0% (HPLC)

Synonym(s):

1,8-Bis(aminomethyl)anthracene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
9061081
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NB.61

Quality Level

Assay

≥98.0% (HPLC)

SMILES string

NCC1=C2C(C=C(C=CC=C3CN)C3=C2)=CC=C1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service