Skip to Content
Merck
All Photos(1)

Key Documents

10039

Sigma-Aldrich

Amoxicillin

≥97.0% (dried material, NT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19N3O5S
CAS Number:
Molecular Weight:
365.40
Beilstein:
7507120
EC Number:
MDL number:
UNSPSC Code:
51101500

Pricing and availability is not currently available.

Assay

≥97.0% (dried material, NT)

impurities

~15% water

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccc(O)cc3)C(=O)N2[C@H]1C(O)=O

InChI

1S/C16H19N3O5S/c1-16(2)11(15(23)24)19-13(22)10(14(19)25-16)18-12(21)9(17)7-3-5-8(20)6-4-7/h3-6,9-11,14,20H,17H2,1-2H3,(H,18,21)(H,23,24)/t9-,10-,11+,14-/m1/s1

InChI key

LSQZJLSUYDQPKJ-NJBDSQKTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

László Szabó et al.
Free radical research, 50(1), 26-38 (2015-10-03)
Penicillins, as bactericidal antibiotics, have been widely used to treat infections for several decades. Their structure contains both aromatic and thioether moieties susceptible to free radical oxidation. The (•)OH induced oxidation mechanism of amoxicillin was investigated by pulse radiolysis techniques
Allison R Bechard et al.
Behavioral neuroscience, 133(2), 247-254 (2019-02-05)
Research using the cocaine self-administration and reinstatement animal model of relapse finds that the beta-lactam antibiotic, ceftriaxone, attenuates cocaine-primed reinstatement of cocaine seeking and upregulates two proteins that regulate glutamate release and reuptake (xCT and GLT-1, respectively) in the nucleus
Thao Thi Nguyen et al.
Scientific reports, 9(1), 9349-9349 (2019-06-30)
This study investigates the photocatalytic degradation of amoxicillin (AMO) by simulated solar irradiation using WO3 as a catalyst. A three-factor-three-level Box-Behnken design (BBD) consisting of 30 experimental runs is employed with three independent variables: initial AMO concentration, catalyst dosage, and
Jakob Menz et al.
Environmental science & technology, 52(11), 6555-6564 (2018-04-10)
There are growing concerns that antibiotic pollution impacts environmental microbiota and facilitates the propagation of antibiotic resistance. However, the prediction or analytical determination of bioavailable concentrations of antibiotics in soil is still subject to great uncertainty. Biological assays are increasingly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service