Skip to Content
Merck
All Photos(1)

Key Documents

03718

Supelco

Glutaraldehyde bis-(O-pentafluorophenylmethyloxime)

analytical standard

Synonym(s):

Glutaraldehyde-O-2,3,4,5,6-PFBHA-Oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H12F10N2O2
CAS Number:
Molecular Weight:
490.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

storage temp.

2-8°C

SMILES string

Fc1c(F)c(F)c(CO\N=C\CCC\C=N\OCc2c(F)c(F)c(F)c(F)c2F)c(F)c1F

InChI

1S/C19H12F10N2O2/c20-10-8(11(21)15(25)18(28)14(10)24)6-32-30-4-2-1-3-5-31-33-7-9-12(22)16(26)19(29)17(27)13(9)23/h4-5H,1-3,6-7H2/b30-4+,31-5+

InChI key

YAGVMVPKFZBARN-IBLZLIEZSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service