Skip to Content
Merck
All Photos(1)

Documents

40-0201

Sigma-Aldrich

Potassium sodium phosphate buffer solution

pH 6.4, 1/15 M

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700

form

liquid

availability

available only in Japan

concentration

1/15 M

pH

6.4

Looking for similar products? Visit Product Comparison Guide

Suitability

Giemsa staining

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service