Skip to Content
Merck
All Photos(1)

Documents

40-0115

Sigma-Aldrich

pH standard solution

pH 4.01 (25 °C), JCSS

Synonym(s):

pH 4.01 Buffer Calibration Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700

Agency

JCSS

availability

available only in Japan

pH

4.01 (25 °C)

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

Application


  • Production of radioisotopes under GMP compliance: Utilization of pH standard solutions in the preparation of [(211)At]NaAt solutions, highlighting the critical role of pH accuracy control in the synthesis of radioisotopes for clinical trials. This application demonstrates the necessity of pH standard solutions in maintaining rigorous quality standards in pharmaceutical and biotechnological research (Naka et al., 2024).

  • Enhancement of environmental cleanup technologies: Research employing pH buffer solutions to stabilize monodispersed and organically modified La(OH)₃ nanocrystals for advanced phosphate removal. This study emphasizes the importance of pH standard solutions in environmental science, particularly for controlling reaction environments in material synthesis (Hu et al., 2024).

  • Improvement of diagnostic accuracy in oncology: A study validating a six-gene signature for non-small-cell lung cancer, where pH buffer solutions were essential for ensuring the stability and accuracy of biomolecular reactions during gene expression analysis. This application showcases the critical role of pH standard solutions in enhancing the reliability of diagnostic tests in medical research (Charkiewicz et al., 2024).

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service