Skip to Content
Merck
All Photos(1)

Key Documents

21-1360

Sigma-Aldrich

Nickel(II) sulfate hexahydrate

JIS special grade, 99.0-102.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiSO4 · 6H2O
CAS Number:
Molecular Weight:
262.85
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
99.0-102.0%
grade:
JIS special grade
form:
crystalline

grade

JIS special grade

vapor density

2.07 (vs air)

Assay

99.0-102.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: nickel

availability

available only in Japan

SMILES string

O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

InChI

1S/Ni.H2O4S.6H2O/c;1-5(2,3)4;;;;;;/h;(H2,1,2,3,4);6*1H2/q+2;;;;;;;/p-2

InChI key

RRIWRJBSCGCBID-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica Mattarozzi et al.
Micron (Oxford, England : 1993), 75, 18-26 (2015-05-20)
Environmental scanning electron microscopy (ESEM) permits to analyze samples in their native-hydrated state, allowing a broad spectrum of biological applications. In this study, ESEM equipped with energy dispersive X-ray spectrometer (EDS) was used as a fast method to analyze tissue
K Amudha et al.
Chemico-biological interactions, 193(1), 57-64 (2011-05-24)
This study was conducted to investigate the beneficial role of naringin on nickel induced nephrotoxicity. Nickel (Ni) (20mg/kg body weight (b.w.) was administered intraperitoneally (i.p.) for 20 days. Naringin was administered orally (20, 40 and 80 mg/kg b.w.) with i.p.
A Tammaro et al.
International journal of immunopathology and pharmacology, 25(2), 519-521 (2012-06-16)
Nickel deficiency leads to reduced iron content in organs and to reduced hemoglobin levels and haematocrit counts. We carried out a clinical trial of oral hyposensitization therapy with low doses of nickel on a group of 20 patients affected by
Xiaohua Xu et al.
Particle and fibre toxicology, 9, 40-40 (2012-11-07)
It has been well recognized that toxicity of fine ambient air particulate matter (PM(2.5)) may depend on its chemical constituents, including components such as soluble metals that may theoretically exert distinctive effects. We have recently demonstrated an important effect of
Charlotte G Mortz et al.
Pediatric allergy and immunology : official publication of the European Society of Pediatric Allergy and Immunology, 24(1), 84-87 (2013-01-22)
We have previously reported patch test reactivity to nickel sulphate in a cohort of unselected infants tested repeatedly at 3-18 months of age. A reproducible positive reaction at 12 and 18 months was selected as a sign of nickel sensitivity provided a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service