Skip to Content
Merck
All Photos(1)

Key Documents

TX0770

Supelco

p-Toluenesulfonic Acid, Monohydrate

GR ACS

Synonym(s):

p-Toluenesulfonic Acid, Monohydrate, 1-Hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O3S · xH2O
CAS Number:
Molecular Weight:
172.20 (anhydrous basis)
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-180-0
grade:
ACS reagent
form:
crystals
solubility:
750 g/L

grade

ACS reagent

Quality Level

Assay

≥99.0% anhydrous basis (acidimetric)

form

crystals

composition

Water, 9.5-11.5%

ign. residue

≤0.1% (600 °C)

color

off-white

pH

1 (20 °C, 650 g/L in H2O, anhydrous substance)

bp

140 °C/27 hPa

mp

56 °C (Elimination of water of crystallisation)

transition temp

flash point 180 °C

solubility

750 g/L

anion traces

sulfate (SO42-): ≤0.3%

cation traces

Fe: ≤0.01%
Na: ≤0.002%
heavy metals (as Pb): ≤0.001%

shipped in

ambient

storage temp.

room temp

InChI

1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2

InChI key

KJIFKLIQANRMOU-UHFFFAOYSA-N

Application

GR ACS

Analysis Note

Appearance (description)hygroscopic crystalsAppearance of solutionconformsAssay (acidimetric; calc. on anhydrous substance)99.0% minColoroff-whiteHeavy metals (as Pb)0.001% maxIron (Fe)0.01% maxResidue after ignition (600°C)0.1% maxSodium (Na)0.002% maxSulfate (SO4)0.3% maxWater (H2O)9.5-11.5%

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service