TANKFIX01
Wall Mounting Bracket for 30 / 60 L Polyethylene Storage Tanks
Save lab bench space
About This Item
Recommended Products
feature
Save lab bench space
packaging
pkg of 1 unit
compatibility
for use with Direct-Q® UV
for use with Elix® Essential UV
for use with Elix® Essential Water Purification Systems
for use with Milli-Q® Direct Water Purification Systems
for use with RiOs™ Essential Water Purification Systems
for use with RiOs™ Water Purification System
shipped in
ambient
Related Categories
General description
Application
Legal Information
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service