Skip to Content
Merck
All Photos(1)

Documents

SNAP2FRMN02

Millipore

SNAP id® 2.0 Mini Blot Holding Frames (double pack)

The new Mini Blot Holders ensure the membrane is horizontal allowing uniform Antibody distribution. The flat profile alows for compact space allocation for storage, shipping & waste management.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.22

manufacturer/tradename

SNAP id®

technique(s)

western blot: suitable

compatibility

for use with Commercially available blocking reagents
for use with Luminata Western HRP Substrates
for use with Nitrocellulose
for use with PVDF (Immobilon membranes)
for use with blØk<TMSYMBOL></TMSYMBOL>-CH Buffer (cat. no. WBAVDCH01)
for use with blØk<TMSYMBOL></TMSYMBOL>-FL Buffer (cat. no. WBAVDFL01)
for use with blØk<TMSYMBOL></TMSYMBOL>-PO Buffer (cat. no. WBAVDP001)
for use with commercially available detection reagents

detection method

chemiluminescent
colorimetric
fluorometric

shipped in

ambient

storage temp.

room temp

General description

The new Mini Blot Holding Frames are stackable and have clear lids to retain humidity. They have an easy locking latch and an umbrella valve for offline incubation.

Application

For use on immunoblots.
The new Mini Blot Holders ensure the membrane is horizontal allowing uniform Antibody distribution. The flat profile alows for compact space allocation for storage, shipping & waste management.

Legal Information

SNAP id is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service