Skip to Content
Merck
All Photos(5)

Key Documents

SCC453

Sigma-Aldrich

OSUMMER.9 Mouse NRAS-Mutant Melanoma Cell Line

Mouse

Synonym(s):

Mouse melanoma cell line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
Product SCC453 is not currently sold in your country. Contact Technical Service

Product Name

OSUMMER.9 Mouse NRAS-Mutant Melanoma Cell Line,

biological source

mouse

Quality Level

packaging

vial of ≥1X10⁶ cells/vial vial

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell culture | mammalian: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

Application

Each vial contains >1X106 viable cells.

Cells are tested negative for infectious diseases by a Mouse Essential CLEAR Panel and have been found to be negative for all pathologies in this test.

Cells are verified to be of mouse origin and negative for interspecies contamination from rat, human, Chinese hamster, Golden Syrian hamster, and non-human primate (NHP) as assessed by a Contamination Clear panel.

Cells are negative for mycoplasma contamination.

Cells are genotyped by STR analysis to verify the unique identity of the cell line.
The OSUMMER (Ohio State University and Moffitt Melanoma Exposed to Radiation) cell lines fulfill a previously unfilled gap in melanoma biology. These cell lines are syngeneic with C57BL/6 laboratory mice and have genetic profiles that are similar to those of human tumors, making them responsive to immunotherapy treatments. These NRAS-mutant cell lines can be used to discover and address potential immunotherapies in NRAS-mutant human melanomas, which make up an estimated 15-30% of human melanomas. These cell lines also enable in vivo testing of immunotherapies with a mouse model.

OSUMMER.6 NRAS mouse melanoma cell line was derived from a male TN mouse (homozygous for the LSL-NrasQ61R allele) exposed to UVA and UVB radiation. OSUMMER.4 can form tumors in vivo without Matrigel injection. WES (whole exome sequencing) revealed high SNV (Single Nucleotide variants) and low CNA (copy number alterations) burden, as well as the presence of UV signature mutations. OSUMMER.4 cells express the melanoma markers PMEL and MelanA and exhibit robust growth in culture. OSUMMER.6 cells demonstrate a relatively low (<lt/>200) mutation burden compared with other OSUMMER NRAS Q61R mutants, and are resistant to MEK inhibition (MEKi).

Source
OSUMMER.6 was derived from melanoma induced in TN (Tyr::CreER) mice exposed to UVA or UVB radiation on postnatal day 3 (1).

References
1. Bowman, R. L., et al. (2021). UVB mutagenesis differs in Nras-and Braf-mutant mouse models of melanoma. Life Science Alliance, 4(9).

Features and Benefits

The OSUMMER.9 melanoma cell line carries the NRAS mutation Q61L and may be used to create syngeneic mouse melanoma models.

Storage and Stability

Cells should be stored in liquid nitrogen. Cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service