Skip to Content
Merck
All Photos(2)

Key Documents

QTUM000EX-M

Quantum® Polishing Cartridge

For Milli-Q systems. Contains Organex & Ionex Resins for trace levels of organics and ions.

Synonym(s):

Quantum® EX Polishing Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.14

packaging

pkg of 1 unit

Looking for similar products? Visit Product Comparison Guide

General description

For Milli-Q systems. Contains Organex & Ionex Resins for trace levels of organics and ions.

Application

For Milli-Q systems. Contains Organex & Ionex Resins for trace levels of organics and ions.

Legal Information

QUANTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service