MABS1931
Anti-CD368/CLEC4D Antibody, clone WEN42
Synonym(s):
C-type lectin domain family 4 member D (UniProt: Q69FH1, also known as C-type lectin superfamily member 8
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
WEN42, monoclonal
mol wt
observed mol wt ~55 kDa
purified by
using protein G
species reactivity
rat
packaging
antibody small pack of 100
technique(s)
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG2a
epitope sequence
Extracellular domain
Protein ID accession no.
UniProt accession no.
storage temp.
2-8°C
Gene Information
rat ... Clec4d(362432)
Specificity
Immunogen
Application
Evaluated by Western Blotting with recombinant Rat CD368/CLEC4D protein.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected recombinant fragment of Rat CD368/CLEC4D expressed with Fc region of human IgG1.
Tested Applications
Immunoprecipitation Analysis: A representative lot immunoprecipitated CD368/CLEC4D in Immunoprecipitation applications (Lobato-Pascual, A., et al. (2013). PLoS One. 8(2):e57406; Lobato-Pascual, A., et al. (2013). Eur J Immunol. 43(12):3167-74).
Immunohistochemistry Applications: A representative lot detected CD368/CLEC4D in Immunohistochemistry applications (Lobato-Pascual, A., et al. (2013). PLoS One. 8(2):e57406).
Western Blotting Analysis: A representative lot detected CD368/CLEC4D in Western Blotting applications (Lobato-Pascual, A., et al. (2013). PLoS One. 8(2):e57406; Lobato-Pascual, A., et al. (2013). Eur J Immunol. 43(12):3167-74).
Flow Cytometry Analysis: A representative lot detected CD368/CLEC4D in Flow Cytometry applications (Lobato-Pascual, A., et al. (2013). PLoS One. 8(2):e57406; Lobato-Pascual, A., et al. (2013). Eur J Immunol. 43(12):3167-74).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service