MABN609
Anti-OCRL/INPP5b Antibody, clone N166A/26
clone N166A/26, from mouse
Synonym(s):
Inositol polyphosphate 5-phosphatase OCRL-1, Lowe oculocerebrorenal syndrome protein
About This Item
biological source
mouse
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
N166A/26, monoclonal
species reactivity
human, rat
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... OCRL(4952)
General description
Specificity
Immunogen
Application
Neuroscience
Developmental Signaling
Western Blotting Analysis: A representative lot detected OCRL/INPP5b in COS cell lysates transiently transfected with GFP-tagged OCRL or RFP-tagged INPP5b, and demonstrated a loss of signal in COS cell lysates transiently transfected with untagged Kv2.1 plasmids (Prof. J. Trimmer, University of California, Davis.).
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected OCRL/INPP5b in human cerebellum and human cardiac myocytes.
Quality
Western Blotting Analysis: 0.5 µg/mL of this antibody detected OCRL/INPP5b in 10 µg of human brain tissue lysate.
Target description
Physical form
Storage and Stability
Analysis Note
Rat brain tissue lysate
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service