Skip to Content
Merck
All Photos(1)

Key Documents

MAB8671F-5

Sigma-Aldrich

Anti-Influenza B Antibody, Core, clone 22D5-12-13

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
HUF 713,000.00

HUF 713,000.00


Estimated to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
500 μG
HUF 713,000.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

HUF 713,000.00


Estimated to ship onApril 07, 2025Details


Request a Bulk Order

conjugate

FITC

Quality Level

clone

22D5-12-13
monoclonal

species reactivity

mouse, human

concentration

(Please refer to the Certificate of Analysis for the lot-specific concentration.)

technique(s)

immunofluorescence: suitable

isotype

IgG2a

Specificity

Influenza B, anti-core specific; reacts with all strains of Influenza B tested. No reactivity with Flu A or other respiratory viruses

Application

Epitope: Core

Physical form

Purified, FITC conjugated. Liquid in 0.02M phosphate buffer, 0.25M sodium chloride, pH 7.6 with 15 mg/mL BSA and 0.1% sodium azide

Storage and Stability

Store at 2° to 8°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service