Skip to Content
Merck
All Photos(1)

Documents

HF000MC100

Millipore

Laminated Card with release liners

No membrane, filter W × L 6 cm × 30 cm, pkg of 100 sheets

Synonym(s):

Plain laminated cards without membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
NACRES:
NB.24

Quality Level

packaging

pkg of 100 sheets

filter W × L

6 cm × 30 cm

Related Categories

General description

Lateral flow tests faciliate quick and convenient detection / quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine. This pack contains 100 laminated cards with release liners for use in lateral flow test manufacturing. Note: this product does not contain Hi-Flow Plus membrane.

Application

Millipore laminated cards are suitable for use in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs, etc.), as well as in women’s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing, and animal health.

Features and Benefits

  • Precut for convenience.
  • Quick delivery and lead time.

Legal Information

Hi-Flow is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service