GF303
G-CSF Protein, Human Recombinant Animal Free
The GCSF protein is a pleiotropic cytokine best known for its specific effects on the proliferation, differentiation & activation of hematopoietic cells of the neutrophilic granulocyte lineage. Manufactured using all non-animal reagents.
Synonym(s):
GCSF, Granulocyte Colony Stimulating Factor, Pluripoietin, Filgrastim, Lenograstim
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
>98% (SDS-PAGE and HPLC)
impurities
<0.1 μg/mg protein endotoxin (<1EU/μg)
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Related Categories
General description
Granulocyte Colony-Stimulating Factor (GCSF) is a pleiotropic cytokine best known for its specific effects on the proliferation, differentiation, and activation of hematopoietic cells of the neutrophilic granulocyte lineage. It is produced mainly by monocytes and macrophages upon activation by endotoxin, TNFα and IFNγ. Other cell types including fibroblasts endothelial cells, astrocytes and bone marrow stromal cells can also secrete GCSF after LPS, IL1 or TNFα activation. In addition, various carcinoma cell lines and myeloblastic leukemia cells can express GCSF constitutively. G-CSF is being used clinically to facilitate hematopoietic recovery after bone marrow transplantation. Human and murine G-CSF are cross-species reactive. Recombinant human G-CSF is an 18.7 kDa protein consisting of 174 amino acid residues.
Product Source: E. coli
Specificity
Cross Reactivty
Mouse
Mouse
Application
Research Category
Inflammation & Immunology
Inflammation & Immunology
Research Sub Category
Growth Factors & Receptors
Growth Factors & Receptors
Packaging
10 ug
Quality
Bioactivity assay: Determined by its ability to stimulate the proliferation of mouse M-NFS-60 cells. The expected ED50 is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.
Physical form
Product is filtered through a 0.2 micron filter before lyophilization.
Recombinant animal free human G-CSF is manufactured using all non-animal reagents
Storage and Stability
Store at -20°C for up to 6 months from date of receipt.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service