Skip to Content
Merck
All Photos(1)

Documents

EZCCP-47K

Millipore

Canine C-peptide ELISA Kit

measures and quantifies C-Peptide in 20 μL serum or plasma

Synonym(s):

C-peptide ELISA, Canine C-peptide Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

product name

Canine C-peptide ELISA,

species reactivity

canine

Quality Level

packaging

kit of 1 × 96 wells

parameter

20 μL sample volume (4hr competitive assay)

technique(s)

ELISA: suitable

input

sample type plasma (K2 EDTA)
sample type serum

NCBI accession no.

UniProt accession no.

application(s)

research use

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 0.24 ng/mL
standard curve range: 0.2-10 ng/mL
accuracy: 95%
linearity: 96-118%

detection method

colorimetric (450nm/590nm)

shipped in

wet ice

storage temp.

2-8°C

General description

C-peptide is a byproduct of normal insulin production by the beta cells in the pancreas and correlates directly to endogenous concentrations of insulin.

Application

3 hour assay, room temperature 20 μL sample size
Research Category
Metabolism
Research Sub Category
Obesity|Metabolic Disorders
This Canine C-Peptide ELISA is used to measure & quantify C-Peptide levels in Metabolism & Endocrine research.
Used to detect/quantify:C-Peptide

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service