Skip to Content
Merck
All Photos(1)

Documents

CDRCHKIT4

Milli-Q

Reverse Osmosis Cartridge

For water purification by reverse osmosis (RO)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104214
eCl@ss:
33050190
NACRES:
JA.14

feature

removes 95%-99% of contaminants including ions, particulates, bacteria, and organic molecules

packaging

pkg of 1 unit

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The Reverse Osmosis membrane removes 95%-99% of contaminants including ions, particulates, bacteria, and organic molecules.

Application

Water Purification by Reverse Osmosis (RO)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service