AP202J
Goat Anti-Rat light chain Antibody, Cy2 conjugate
Chemicon®, from goat
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
goat
Quality Level
conjugate
CY2 conjugate
antibody form
affinity purified immunoglobulin
antibody product type
secondary antibodies
clone
polyclonal
species reactivity
rat
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
western blot: suitable
isotype
IgG
shipped in
wet ice
target post-translational modification
unmodified
Related Categories
General description
Light chains covalently bound to a pair of heavy chains to form an antibody molecule. Immunoglobulin light chains occur in two types, kappa or lambda. Light chain′s mass is about 25,000 Daltons per mole. Each light chain consists of about 250 amino-acids long. These are usually designated by the Greek letters kappa and lambda. The ratio of kappa to lambda found in the immunoglobulin population varies by species.
Specificity
Minimal cross-reaction with bovine, goat, horse, human, mouse, rabbit, and sheep.
Application
Goat anti-Rat light chain Antibody, Cy2 conjugate detects level of light chain & has been published & validated for use in ELISA & WB.
Western Blotting: 1:50 -1:200
ELISA: 1:50 -1:200
Optimal working dilutions must be determined by the end user.
ELISA: 1:50 -1:200
Optimal working dilutions must be determined by the end user.
Target description
25 kDa
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service