Skip to Content
Merck
All Photos(2)

Documents

ABN343

Sigma-Aldrich

Anti-NIPSNAP Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Non-neuronal SNAP25-like protein, NIPSNAP1, NIPSNAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NIPSNAP1(8508)

General description

NIPSNAP is a member of an evolutionarily well conserved gene family and has a strong sequence similarity to the central portion of a protein encoded by C. elegans chromosome III between a 4-nitrophenylphosphatase (NIP) domain and non-neuronal SNAP25-like protein. Recent studies have indicated that NIPSNAP is involved in the regulation of the Ca2+-selective transient receptor potential vanilloid channel 6 (TRPV6). NIPSNAP1 associates with TRPV6 at the plasma membrane and inhibits TRPV6 currents. Other studies show that NIPSNAP’s expression is reduced in the phenylketonuria (PKU) brain, suggesting that NIPSNAP may play a role in memory.

Immunogen

KLH-conjugated linear peptide corresponding to human NIPSNAP.

Application

Detect the NIPSNAP protein using this Anti-NIPSNAP validated for use in WB & IHC.
Immunohistochemistry Analysis: 2.5 µg/mL of this antibody from a representative lot detected NIPSNAP in human brain tissue.

Quality

Evaluated by Western Blot in human brain tissue lysate.

Western Bloting Analysis: 1 µg/mL of this antibody detected NIPSNAP in 15 µg of human brain tissue lysate.

Target description

~33 kDa observed

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service