Skip to Content
Merck
All Photos(1)

Documents

8.41456

Sigma-Aldrich

DL-Camphor

for synthesis

Synonym(s):

DL-Camphor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
MDL number:
UNSPSC Code:
12352115
EC Index Number:
200-945-0
NACRES:
NA.22

vapor pressure

1.2 hPa ( 20 °C)

Quality Level

Assay

≥95.0% (GC)

form

solid

autoignition temp.

460 °C

potency

>5000 mg/kg LD50, oral (Rat)

expl. lim.

0.6-4.5 % (v/v)

bp

205 °C/1013 hPa

mp

180 °C

transition temp

flash point 66 °C

solubility

1.5 g/L

density

0.99 g/cm3 at 20 °C

bulk density

800 kg/m3

storage temp.

2-30°C

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3

InChI key

DSSYKIVIOFKYAU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Melting range (lower value): ≥ 170 °C
Melting range (upper value): ≤ 176 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 2 Inhalation

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

147.9 °F - closed cup

Flash Point(C)

64.4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service