Skip to Content
Merck
All Photos(1)

Documents

5.08940

Millipore

G 418 Sulfate, Sterile-Filtered Solution in 100 mM HEPES, Cell Culture Tested

Synonym(s):

G 418 Sulfate, Sterile-Filtered Solution in 100 mM HEPES, Cell Culture Tested, G418, G 418

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H40N4O10 · 2H2SO4
CAS Number:
Molecular Weight:
692.71
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (TLC)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

storage temp.

−20°C

InChI

1S/C20H40N4O10.2H2O4S/c1-6(25)14-11(27)10(26)9(23)18(32-14)33-15-7(21)4-8(22)16(12(15)28)34-19-13(29)17(24-3)20(2,30)5-31-19;2*1-5(2,3)4/h6-19,24-30H,4-5,21-23H2,1-3H3;2*(H2,1,2,3,4)/t6?,7-,8+,9+,10+,11-,12-,13-,14+,15+,16-,17-,18+,19-,20+;;/m0../s1

InChI key

UHEPSJJJMTWUCP-NKCAIAFTSA-N

General description

Recommended reaction conditions: The optimal concentration of G418 for selection of resistance will vary according to the organism and/or cell type under investigation. In general, the concentration of active drug required for selection is as follows: Dictyostelium sp.: 10-100 µg/ml Plant cells: 10-100 µg/ml Yeast cells: 0.5-1.0 µg/ml Mammalian cells: 0.1-2.0 µg/ml

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Physical form

Sterile-filtered solution of Cat. No. 345810 supplied at 50 mg/ml in 100 mM HEPES, pH 7.3.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 1 year at -20°C.

Other Notes

Ethier, S.P., and Taback, E. 1993. Cancer Lett.74, 189.
Santerre, R.F., et al. 1991. Methods Mol. Biol.7, 245.
Maniatis, T., et al. 1989. In Molecular Cloning, A Laboratory Manual, Second Edition, Cold Spring Harbor, NY.
Edwards, S.A., and Adamson, E.D. 1987. J. Cell Physiol.133, 46.
Ernst, J.F., and Chan, R.K. 1985. J. Bacteriol.163, 8.
Canaani, D., and Berg, P. 1982. Proc. Natl. Acad. Sci. USA79, 5166.
Hirth, K.P., et al. 1982. Proc. Natl. Acad. Sci. USA79, 7356.
Ursic, D., et al. 1981. Biochem. Biophys. Res. Commun.101, 1031.
Jimenez, A., and Davies, J. 1980. Nature287, 869.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service