Skip to Content
Merck
All Photos(1)

Documents

482200

Sigma-Aldrich

Nimodipine

An L-type Ca2+ channel blocker.

Synonym(s):

Nimodipine, 1,4-Dihydro-2,6-dimethyl-4-(3ʹ-nitrophenyl)-3,5-pyridinedicarboxylic acid 2-methoxyethyl-1-methylethyl Ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2O7
CAS Number:
Molecular Weight:
418.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

>98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 25 mg/mL
methanol: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C21H26N2O7/c1-12(2)30-21(25)18-14(4)22-13(3)17(20(24)29-10-9-28-5)19(18)15-7-6-8-16(11-15)23(26)27/h6-8,11-12,19,22H,9-10H2,1-5H3

InChI key

UIAGMCDKSXEBJQ-UHFFFAOYSA-N

General description

An L-type Ca2+ channel blocker. Enhances memory in old or brain-damaged animals. Ameliorates experimental diabetic neuropathy in streptozotocin-induced diabetic rats.
An L-type Ca2+ channel blocker. Enhances memory in old or brain-damaged animals. Ameliorates experimental diabetic neuropathy in streptozotocin-induced diabetic rats. Also reported to block TNF-α and LPS-induced iNOS activity (IC50 = 36 pM) in mouse astroglial cells.

Biochem/physiol Actions

Cell permeable: no
Primary Target
L-type Ca2+ channel blocker
Product does not compete with ATP.
Reversible: no
Target IC50: 36 pM blocking TNF-α and LPS-induced iNOS activity in mouse astroglial cells

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhu, D., et al. 1999. Life. Sci.65, PL 221.
Marchetti, C. and Usai, C. 1996. Neurosci. Lett. 207, 77.
Kappelle, A.C., et al. 1994. Br. J. Pharmacol.111, 887.
Weiss, J.H., et al. 1994. J. Neurochem. 62, 372.
Kappelle, A.C., et al. 1993. Br. J. Pharmacol.108, 780.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ariel Caviedes et al.
Cell death & disease, 12(1), 4-4 (2021-01-09)
Cell death by glutamate excitotoxicity, mediated by N-methyl-D-aspartate (NMDA) receptors, negatively impacts brain function, including but not limited to hippocampal neurons. The NF-κB transcription factor (composed mainly of p65/p50 subunits) contributes to neuronal death in excitotoxicity, while its inhibition should

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service