Skip to Content
Merck
All Photos(1)

Key Documents

371545

Sigma-Aldrich

Guanosine 5ʹ-O-(3-Thiotriphosphate), Tetralithium Salt - CAS 94825-44-2 - Calbiochem

G-protein-activating GTP analog, which is more resistant to hydrolysis than GTP.

Synonym(s):

Guanosine 5ʹ-O-(3-Thiotriphosphate), Tetralithium Salt - CAS 94825-44-2 - Calbiochem, GTPγ-S, 4Li

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N5O13P3S · 4Li
CAS Number:
Molecular Weight:
562.98
MDL number:
UNSPSC Code:
12352200

Assay

≥85% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

impurities

≤12% GDP

color

white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

G-protein-activating GTP analog that is more resistant to hydrolysis than GTP. Has a direct stimulatory effect on Ins(1,4,5)P3 formation in membranes.
G-protein-activating GTP analog, which is more resistant to hydrolysis than GTP. Induces exocytosis in isolated mouse pancreatic acinar cells. Also reported to stimulate insulin release from pancreatic β-cells.

Biochem/physiol Actions

Cell permeable: no
Primary Target
G-protein-activating GTP analog
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions lose about 5% of their activity after 6 months at -20°C.

Other Notes

Iino, S., et al. 2000. FEBS Lett.479, 46.
Martelli, A.M., et al. 1996. Biochem. Biophys. Res. Commun. 218, 182.
Hess, S.D., et al. 1993. Science 259, 1169.
Ekstrom, R.C., et al. 1992. J. Biol. Chem.267, 22183.
McGuirk, S.M., and Dolphin, A.C. 1992. Neurosci.49, 117.
Vaca, L., et al. 1992. Eur. J. Physiol.422, 66.
Silk, S.T., et al. 1989. J. Biol. Chem.264, 21466.
Gilman, A.G. 1987. Annu. Rev. Biochem.56, 615.
Orellana, S., et al. 1987. J. Biol. Chem..262, 1638.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service