Skip to Content
Merck
All Photos(1)

Key Documents

17-10459

Sigma-Aldrich

Magna ChIP® HT96 ChIP Plate Set

The Magna ChIP HT96 ChIP Plate se allows running of partial plates to minimize the risk of cross contamination of samples in sensitive endpoint analyses, such as qPCR.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NA.32

Quality Level

sterility

non-sterile

manufacturer/tradename

Magna ChIP®
Upstate®

technique(s)

ChIP: suitable
immunoprecipitation (IP): suitable

plate size

96 wells

shipped in

ambient

Related Categories

General description

The ChIP plate is a clear, sterile polystyrene 96-well plate with conical bottoms. Individual wells will accommodate up to 250 µL. It is provided with three plates seals and is fully compatible with automated platforms.

Application

The Magna ChIP HT96 ChIP Plate set is intended for use with the Magna ChIP HT96 kit. This plate allows running of partial plates to minimize the risk of cross contamination of samples in sensitive endpoint analyses, such as qPCR. See the HT96 kit protocol (Cat. No. 17-10077) for specific use instructions.

Components

2 x 96-well ChIP Plates

3 x Plate Seals

Physical form

Color Code: Clear

Other Notes

Automation Compatible: No

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service